Merck
  • Controlled reversible assembly of gold nanoparticles as a new colorimetric and sensitive detection of glucose-6-phosphate dehydrogenase deficiency.
Controlled reversible assembly of gold nanoparticles as a new colorimetric and sensitive detection of glucose-6-phosphate dehydrogenase deficiency.
Analytica chimica acta (2020-06-07)
Usa Boonyuen, Aun Praoparotai, Kamonwan Chamchoy, Thitiluck Swangsri, Chompunuch Warakulwit, Teeraporn Suteewong
ABSTRACT
Recently, several studies have examined possible applications of nanoparticles for the development of electronic and optical sensors. The plasmon absorbance of gold nanoparticles has been used extensively to study biomolecular processes, including nicotinamide adenine dinucleotide/nicotinamide adenine dinucleotide phosphate-dependent enzymatic reactions. In this report, we describe the development of gold nanoparticles as a new colorimetric and sensitive detection method of glucose-6-phosphate dehydrogenase deficiency by means of controlled reversible assembly of gold nanoparticles. 3-nm polyvinylpyrrolidone/N,N'-dimethylaminopyridine-stabilized gold nanoparticles were synthesized, characterized and applied for an in vitro activity assay of 11 recombinant human glucose-6-phosphate dehydrogenase variants. Differences in the activity of the glucose-6-phosphate dehydrogenase variants from different deficiency classes were readily detected using the synthesized gold nanoparticles. The developed method can be easily distinguished with color change by naked eye for the detection of glucose-6-phosphate dehydrogenase deficiency. Moreover, we are the first to propose the segregation mechanism of polyvinylpyrrolidone/N,N'-dimethylaminopyridine-stabilized gold nanoparticles by reduced nicotinamide adenine dinucleotide phosphate. The method enables visual detection of glucose-6-phosphate dehydrogenase deficiency, which could be further developed for diagnostic testing of glucose-6-phosphate dehydrogenase deficiency.