Merck
  • Silencing Sirtuin 6 induces cell cycle arrest and apoptosis in non-small cell lung cancer cell lines.
Silencing Sirtuin 6 induces cell cycle arrest and apoptosis in non-small cell lung cancer cell lines.
Genomics (2020-05-04)
Varunkumar Krishnamoorthy, Ravikumar Vilwanathan
ABSTRACT
Sirtuins (SIRT1-7), are NAD-dependent deacetylases and ADP-ribosyl transferases, plays a major part in carcinogenesis. The previous report suggests that in cancer, sirtuins gained tremendous interest and critical regulators of the unusual processes. In carcinogenesis, sirtuins possess either tumor suppressor or promoter. However, in lung cancer condition the studies of sirtuins are less studied. Hence, this designed study investigates the impact of multifaceted sirtuins in NSCLC cells. We evaluated the mRNA and protein expressions of sirtuins by RTPCR and western blot. We found SIRT6 significantly overexpressed in NCI-H520, A549, and NCI-H460 compared with the normal BEAS-2B cell line. Silencing of SIRT6 by siRNA in NSCLC cells caused activation of p53/p21 mediated inhibition of cell proliferation leading to arrest in cell cycle and apoptosis induction. Our results implied that SIRT6 is a tumor promoter in NSCLC development, progression, and regulation. The silencing of SIRT6 to be a novel therapy for lung cancer.