Merck
  • PTSD is associated with neuroimmune suppression: evidence from PET imaging and postmortem transcriptomic studies.
PTSD is associated with neuroimmune suppression: evidence from PET imaging and postmortem transcriptomic studies.
Nature communications (2020-05-14)
Shivani Bhatt, Ansel T Hillmer, Matthew J Girgenti, Aleksandra Rusowicz, Michael Kapinos, Nabeel Nabulsi, Yiyun Huang, David Matuskey, Gustavo A Angarita, Irina Esterlis, Margaret T Davis, Steven M Southwick, Matthew J Friedman, Ronald S Duman, Richard E Carson, John H Krystal, Robert H Pietrzak, Kelly P Cosgrove
ABSTRACT
Despite well-known peripheral immune activation in posttraumatic stress disorder (PTSD), there are no studies of brain immunologic regulation in individuals with PTSD. [11C]PBR28 Positron Emission Tomography brain imaging of the 18-kDa translocator protein (TSPO), a microglial biomarker, was conducted in 23 individuals with PTSD and 26 healthy individuals-with or without trauma exposure. Prefrontal-limbic TSPO availability in the PTSD group was negatively associated with PTSD symptom severity and was significantly lower than in controls. Higher C-reactive protein levels were also associated with lower prefrontal-limbic TSPO availability and PTSD severity. An independent postmortem study found no differential gene expression in 22 PTSD vs. 22 controls, but showed lower relative expression of TSPO and microglia-associated genes TNFRSF14 and TSPOAP1 in a female PTSD subgroup. These findings suggest that peripheral immune activation in PTSD is associated with deficient brain microglial activation, challenging prevailing hypotheses positing neuroimmune activation as central to stress-related pathophysiology.