Merck
AAV-mediated FOXG1 gene editing in human Rett primary cells.
European journal of human genetics : EJHG (2020-06-17)
Susanna Croci, Miriam Lucia Carriero, Katia Capitani, Sergio Daga, Francesco Donati, Filomena Tiziana Papa, Elisa Frullanti, Diego Lopergolo, Vittoria Lamacchia, Rossella Tita, Annarita Giliberti, Elisa Benetti, Francesca Niccheri, Simone Furini, Caterina Lo Rizzo, Silvestro Giovanni Conticello, Alessandra Renieri, Ilaria Meloni
ABSTRACT
Variations in the Forkhead Box G1 (FOXG1) gene cause FOXG1 syndrome spectrum, including the congenital variant of Rett syndrome, characterized by early onset of regression, Rett-like and jerky movements, and cortical visual impairment. Due to the largely unknown pathophysiological mechanisms downstream the impairment of this transcriptional regulator, a specific treatment is not yet available. Since both haploinsufficiency and hyper-expression of FOXG1 cause diseases in humans, we reasoned that adding a gene under nonnative regulatory sequences would be a risky strategy as opposed to a genome editing approach where the mutated gene is reversed into wild-type. Here, we demonstrate that an adeno-associated viruses (AAVs)-coupled CRISPR/Cas9 system is able to target and correct FOXG1 variants in patient-derived fibroblasts, induced Pluripotent Stem Cells (iPSCs) and iPSC-derived neurons. Variant-specific single-guide RNAs (sgRNAs) and donor DNAs have been selected and cloned together with a mCherry/EGFP reporter system. Specific sgRNA recognition sequences were inserted upstream and downstream Cas9 CDS to allow self-cleavage and inactivation. We demonstrated that AAV serotypes vary in transduction efficiency depending on the target cell type, the best being AAV9 in fibroblasts and iPSC-derived neurons, and AAV2 in iPSCs. Next-generation sequencing (NGS) of mCherry+/EGFP+ transfected cells demonstrated that the mutated alleles were repaired with high efficiency (20-35% reversion) and precision both in terms of allelic discrimination and off-target activity. The genome editing strategy tested in this study has proven to precisely repair FOXG1 and delivery through an AAV9-based system represents a step forward toward the development of a therapy for Rett syndrome.
Sigma-Aldrich
Anti-Pax6 Antibody, ascites fluid, clone AD2.38, Chemicon®