Merck
NF1-RAC1 axis regulates migration of the melanocytic lineage.
Translational oncology (2020-09-07)
Lionel Larribère, Yohanes Cakrapradipta Wibowo, Nitin Patil, Mohammed Abba, Isabel Tundidor, Rubén Gerardo Aguiñón Olivares, Heike Allgayer, Jochen Utikal
ABSTRACT
Metastases's spreading is the main cause of mortality for advanced stage cancer patients, including melanoma. The formation of metastases is favored by enhanced migratory and invasive capacities of tumor cells. Tumor suppressor gene NF1 is a negative regulator of RAS and its deregulation plays an important role in several aspects of melanoma transformation and progression. However, very little is described about the role of NF1 in cellular migration and invasion. In this study, our results show on the one hand, that the loss of NF1 expression delays migration of human melanoblasts via a RAC1-dependent mechanism. On the other hand, our data indicate that NF1 loss in melanoma cells is enhancing migration, intravasation and metastases formation in vivo. Moreover, not only this phenotype is associated with an upregulation of PREX1 but also patient-derived melanoma samples with low NF1 expression present increased levels of PREX1. In sum, our study brings new elements on the mechanism controlling cellular migration in the context of NF1 loss. These data are of prime interest to improve treatment strategies against all NF1-mutated tumors, including this subtype of melanoma.
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture