Merck
  • Gapmer Antisense Oligonucleotides Targeting 5S Ribosomal RNA Can Reduce Mature 5S Ribosomal RNA by Two Mechanisms.
Gapmer Antisense Oligonucleotides Targeting 5S Ribosomal RNA Can Reduce Mature 5S Ribosomal RNA by Two Mechanisms.
Nucleic acid therapeutics (2020-06-27)
Adam J Pollak, Justin H Hickman, Xue-Hai Liang, Stanley T Crooke
ABSTRACT
In this study, we demonstrate that 5S ribosomal RNA (rRNA), a highly structured and protein-bound RNA, is quite difficult to reduce with antisense oligonucleotides (ASOs). However, we found a single accessible site that was targetable with a high-affinity complementary ASO. The ASO appeared to bind to the site, recruit RNaseH1, and cause degradation of the 5S RNA. Intriguingly, we also observed that the same ASO induced an accumulation of pre-5S RNA, which may contribute to reduced levels of mature 5S rRNA. As expected, ASO mediated reduction of 5S RNA, and modest inhibition of processing of pre-5S RNA resulted in nucleolar toxicity. However, the toxicity induced was minimal compared with actinomycin D, consistent with its modest effects on pre-5S rRNA. Mechanistically, we show that the accumulation of pre-5S rRNA required ASO hybridization to the cognate rRNA sequence but was independent of RNaseH1 activity. We found that Ro60 and La, proteins known to bind misprocessed RNAs, likely sequester the ASO-pre-5S rRNA species and block RNaseH1 activity, thus identifying another example of competitive mechanisms mediated by proteins that compete with RNaseH1 for binding to ASO-RNA heteroduplexes.