Merck
  • Green synthesized selenium doped zinc oxide nano-antibiotic: synthesis, characterization and evaluation of antimicrobial, nanotoxicity and teratogenicity potential.
Green synthesized selenium doped zinc oxide nano-antibiotic: synthesis, characterization and evaluation of antimicrobial, nanotoxicity and teratogenicity potential.
Journal of materials chemistry. B (2020-08-20)
Abdul Majeed, Faryal Javed, Sundus Akhtar, Uzma Saleem, Fareeha Anwar, Bashir Ahmad, Akhtar Nadhman, Gul Shahnaz, Irshad Hussain, Syed Zajif Hussain, Muhammad Farhan Sohail
ABSTRACT
A facile, green synthesis of selenium doped zinc oxide nano-antibiotic (Se-ZnO-NAB) using the Curcuma longa extract is reported to combat the increased emergence of methicillin-resistant Staphylococcus aureus (MRSA). The developed Se-ZnO-NAB were characterized for their physicochemical parameters and extensively evaluated for their toxicological potential in an animal model. The prepared Se-ZnO-NABs were characterized via Fourier transformed infrared spectroscopy to get functional insight into their surface chemistry, scanning electron microscopy revealing the polyhedral morphology with a size range of 36 ± 16 nm, having -28.9 ± 6.42 mV zeta potential, and inductively coupled plasma optical emission spectrometry confirming the amount of Se and Zn to be 14.43 and 71.70 mg L-1 respectively. Moreover, the antibacterial activity against MRSA showed significantly low minimum inhibitory concentration at 6.2 μg mL-1 when compared against antibiotics. Also, total protein content and reactive oxygen species production in MRSA, under the stressed environment of Se-ZnO-NAB, significantly (p < 0.05) decreased compared to the negative control. Moreover, the results of acute oral toxicity in rats showed moderate variations in blood biochemistry and histopathology of vital organs. The teratogenicity and fetal evaluations also revealed some signs of toxicity along with changes in biochemical parameters. The overall outcomes suggest that Se-ZnO-NAB can be of significant importance for combating multi-drug resistance but must be used with extreme caution, particularly in pregnancy, as moderate toxicity was observed at a toxic dose of 2000 mg kg-1.