Merck
  • ETV6 germline mutations cause HDAC3/NCOR2 mislocalization and upregulation of interferon response genes.
ETV6 germline mutations cause HDAC3/NCOR2 mislocalization and upregulation of interferon response genes.
JCI insight (2020-08-26)
Marlie H Fisher, Gregory D Kirkpatrick, Brett Stevens, Courtney Jones, Michael Callaghan, Madhvi Rajpurkar, Joy Fulbright, Megan A Cooper, Jesse Rowley, Christopher C Porter, Arthur Gutierrez-Hartmann, Kenneth Jones, Craig Jordan, Eric M Pietras, Jorge Di Paola
ABSTRACT
ETV6 is an ETS family transcription factor that plays a key role in hematopoiesis and megakaryocyte development. Our group and others have identified germline mutations in ETV6 resulting in autosomal dominant thrombocytopenia and predisposition to malignancy; however, molecular mechanisms defining the role of ETV6 in megakaryocyte development have not been well established. Using a combination of molecular, biochemical, and sequencing approaches in patient-derived PBMCs, we demonstrate abnormal cytoplasmic localization of ETV6 and the HDAC3/NCOR2 repressor complex that led to overexpression of HDAC3-regulated interferon response genes. This transcriptional dysregulation was also reflected in patient-derived platelet transcripts and drove aberrant proplatelet formation in megakaryocytes. Our results suggest that aberrant transcription may predispose patients with ETV6 mutations to bone marrow inflammation, dysplasia, and megakaryocyte dysfunction.