Merck
  • The anti-B7-H4 checkpoint synergizes trastuzumab treatment to promote phagocytosis and eradicate breast cancer.
The anti-B7-H4 checkpoint synergizes trastuzumab treatment to promote phagocytosis and eradicate breast cancer.
Neoplasia (New York, N.Y.) (2020-09-24)
Xiaochen Hu, Yiwen Liu, Xiusen Zhang, Dejiu Kong, Jinyu Kong, Di Zhao, Yibo Guo, Lingyun Sun, Luoyi Chu, Shupei Liu, Xurong Hou, Feng Ren, Ying Zhao, Chengbiao Lu, Desheng Zhai, Xiang Yuan
ABSTRACT
Trastuzumab is a humanized mAb used to treat HER2-overexpressing breast cancer; however its mechanisms remain to be fully elucidated. Previous studies suggest a role for immunity in mediating trastuzumab-specific antitumor effects. This study evaluated the role(s) of trastuzumab and other antibodies on macrophage activation and Ab-dependent cell-mediated phagocytosis (ADCP) of HER2+ breast cancer cells in vitro and in vivo. We employed orthotopic implantation of HER2+ murine breast cancer (BC) cells in immunocompetent mouse models, a human HER2+ BC xenograft in an immune humanized mouse model, and human PDXs involving adoptive transfer of autologous macrophages to simulate an endogenous mammary tumor-immune microenvironment. Our study demonstrated that trastuzumab greatly and consistently increased macrophage frequency and tumor-cell phagocytosis, and that concurrent knockdown of B7-H4 by a neutralizing antibody increased immune cell infiltration and promoted an antitumor phenotype. Furthermore, neoadjuvant trastuzumab therapy significantly upregulated B7-H4 in the cancer-infiltrating macrophages of HER2+ BC patients, which predicted poor trastuzumab response. We suggest that strategies to specifically enhance ADCP activity might be critical to overcoming resistance to HER2 mAb therapies by inhibiting tumor growth and potentially enhance antigen presentation. Furthermore, these results advance the understanding of macrophage plasticity by uncovering a dual role for ADCP in macrophages involving elimination of tumors by engulfing cancer cells while causing a concomitant undesired effect by upregulating immunosuppressive checkpoints.