Merck
  • Caspase Recruitment Domain-Containing Protein 9 Expression is a Novel Prognostic Factor for Lung Adenocarcinoma.
Caspase Recruitment Domain-Containing Protein 9 Expression is a Novel Prognostic Factor for Lung Adenocarcinoma.
OncoTargets and therapy (2020-09-29)
Nanako Miwa, Tatsuya Nagano, Naoe Jimbo, Ryota Dokuni, Tatsunori Kiriu, Chihiro Mimura, Yuichiro Yasuda, Masahiro Katsurada, Masatsugu Yamamoto, Motoko Tachihara, Yugo Tanaka, Kazuyuki Kobayashi, Tomoo Itoh, Yoshimasa Maniwa, Yoshihiro Nishimura
ABSTRACT
Caspase recruitment domain-containing protein 9 (CARD9) is expressed at high levels in bone marrow cells and has a crucial role in innate immunity. Current studies indicate that CARD9 also plays a key role in tumor progression, but there are few reports on the role of CARD9 in lung cancer. The aim of this study was to clarify the role of CARD9 in lung adenocarcinoma. Lung adenocarcinoma tumor samples from 74 patients who underwent complete resection at Kobe University Hospital from January 2014 to December 2014 were analyzed by immunohistochemistry. The role of CARD9 in cancer cells was analyzed using lung cancer cell lines treated with CARD9 siRNA. High expression of CARD9 was observed in 32.4% of tumors, and compared to low expression of CARD9, high expression was associated with poorer overall survival (P = 0.0365). Univariate and multivariate analyses showed that high expression of CARD9 was an independent prognostic factor. Knockdown of CARD9 in lung adenocarcinoma cells inhibited proliferation but did not increase apoptosis. In addition, CARD9 activated the NF-κB pathway in a lung adenocarcinoma cell line. CARD9 was shown to be an independent prognostic factor of poor outcome for lung cancer and may represent a molecular target for treatment.