Merck
  • Role of specialized composition of SWI/SNF complexes in prostate cancer lineage plasticity.
Role of specialized composition of SWI/SNF complexes in prostate cancer lineage plasticity.
Nature communications (2020-11-05)
Joanna Cyrta, Anke Augspach, Maria Rosaria De Filippo, Davide Prandi, Phillip Thienger, Matteo Benelli, Victoria Cooley, Rohan Bareja, David Wilkes, Sung-Suk Chae, Paola Cavaliere, Noah Dephoure, Anne-Christine Uldry, Sophie Braga Lagache, Luca Roma, Sandra Cohen, Muriel Jaquet, Laura P Brandt, Mohammed Alshalalfa, Loredana Puca, Andrea Sboner, Felix Feng, Shangqian Wang, Himisha Beltran, Tamara Lotan, Martin Spahn, Marianna Kruithof-de Julio, Yu Chen, Karla V Ballman, Francesca Demichelis, Salvatore Piscuoglio, Mark A Rubin
ABSTRACT
Advanced prostate cancer initially responds to hormonal treatment, but ultimately becomes resistant and requires more potent therapies. One mechanism of resistance observed in around 10-20% of these patients is lineage plasticity, which manifests in a partial or complete small cell or neuroendocrine prostate cancer (NEPC) phenotype. Here, we investigate the role of the mammalian SWI/SNF (mSWI/SNF) chromatin remodeling complex in NEPC. Using large patient datasets, patient-derived organoids and cancer cell lines, we identify mSWI/SNF subunits that are deregulated in NEPC and demonstrate that SMARCA4 (BRG1) overexpression is associated with aggressive disease. We also show that SWI/SNF complexes interact with different lineage-specific factors in NEPC compared to prostate adenocarcinoma. These data point to a role for mSWI/SNF complexes in therapy-related lineage plasticity, which may also be relevant for other solid tumors.
Sigma-Aldrich
Indole-3-acetic acid sodium salt, suitable for plant cell culture, BioReagent, ≥98%