Merck
  • Increased Circulatory Extrarenal 1α,25-Dihydroxyvitamin D3 in Bilaterally Nephrectomized Rats.
Increased Circulatory Extrarenal 1α,25-Dihydroxyvitamin D3 in Bilaterally Nephrectomized Rats.
Endocrine, metabolic & immune disorders drug targets (2020-05-31)
Murat Dabak, Durrin O Dabak, Tuncay Kuloglu, Ersoy Baydar, Hakan Bulut, Ozgur Ozoner
ABSTRACT
Extrarenal 1α,25-dihydroxyvitamin D3 (1,25-D) locally produced by immune cells plays crucial roles in the regulation of the immune system. However, in vivo status of extrarenal 1,25-D and 25-hydroxyvitamin D (25-D) in acute inflammatory conditions are unknown. The aim of this study was to determine the extrarenal 1,25-D level in circulation in bilaterally nephrectomized rats, induced by low-dose lipopolysaccharide (LPS). Renal 1,25-D synthesis was terminated through bilateral nephrectomy in rats. The rats received intraperitoneal LPS (50 μg/kg BW) three times and the experiment was ended 24 hours after nephrectomy. Serum 1,25-D, 25-D, calcium, phosphorus, intact parathyroid hormone, and calcitonin levels were measured and immunohistochemistry was applied to detect the sources of extrarenal 1,25- D synthesis. Circulatory 1,25-D concentration remarkably increased in both LPS-treated and non-treated bilaterally nephrectomized rats. Elevated circulatory 1,25-D did not have hypercalcemic endocrinal effects. The increased 1,25-D level also resulted in a concurrent rapid and dramatic depletion of circulatory 25-D. Extrarenal 1,25-D could enter into the systemic circulation and, therefore, might have systemic effects besides its autocrine and paracrine functions.
Sigma-Aldrich
Phosphate buffered saline, tablet
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, ≥10,000 EU/mg, purified by phenol extraction