Merck
  • Preparation of a Cellulose Column for Enhancing the Sensing Efficiency of the Biocide 2-n-Octyl-4-Isothiazolin-3-One.
Preparation of a Cellulose Column for Enhancing the Sensing Efficiency of the Biocide 2-n-Octyl-4-Isothiazolin-3-One.
Polymers (2020-11-20)
Seong Ho Hong, Sang Wook Kang
ABSTRACT
In this study, a cellulose acetate (CA) membrane with pores generated by a water pressure treatment was investigated for its ability to serve as a pretreatment filter device for the detection of 2-n-octyl-4-isothiazolin-3-one (OIT). Pores were generated by applying a water pressure of 8 bar to a membrane manufactured using a CA-based polymer solution. The CA used for the manufacturing was an environment-friendly, low-cost and highly energy-efficient material. Furthermore, since the fabricated porous CA polymeric film possessed many hydrophilic functional groups, it could strongly bind hydrophilic substances while avoiding interaction with hydrophobic substances. OIT, which comprises a hydrophobic bond that forms weak bonds over time, can break down more easily than hydrophilic impurities. The different extents of interaction occurring between either the toxic fungicide OIT or the hydrophilic impurities and the CA film were determined by Fourier-transform infrared (FT-IR) spectroscopy. The physicochemical changes in the resulting membrane, which occurred when the pores were generated, were investigated through scanning electron microscopy (SEM) and thermogravimetric analysis (TGA).