Merck
  • alpha4 Integrin/FN-CS1 mediated leukocyte adhesion to brain microvascular endothelial cells under flow conditions.
alpha4 Integrin/FN-CS1 mediated leukocyte adhesion to brain microvascular endothelial cells under flow conditions.
Journal of neuroimmunology (2009-04-07)
Shumei Man, Barbara Tucky, Nika Bagheri, Xiaolong Li, Rabina Kochar, Richard M Ransohoff
ABSTRACT
Insights into sequential leukocyte-endothelial interactions during leukocyte trafficking have been obtained through experiments using human umbilical vein endothelial cells (HUVEC) under flow conditions. To investigate leukocyte-brain endothelial cell interactions, we developed a dynamic in vitro system, using Transfected Human Brain Microvascular Endothelial Cells (THBMEC) and a parallel plate flow chamber. Human peripheral blood mononuclear cells (PBMC) were perfused across confluent THBMEC cultures at a velocity that approximates the rate found in human brain capillaries. Leukocyte-THBMEC interactions were visualized by phase-contrast microscopy, and images were captured on a CCD camera. To simulate inflammatory conditions, we activated THBMEC with the inflammatory cytokines tumor necrosis factor alpha (TNF-alpha) and interferon gamma (IFN-gamma), which up-regulated chemokine and adhesion molecule expression in THBMEC without affecting the distribution of immunoreactivity for tight junction-associated proteins. PBMC adhesion was enhanced by cytokine-mediated activation of THBMEC. G protein-coupled receptor (GPCR) activation was essential for leukocyte-THBMEC interaction, as pertussis toxin (PTX) treatment of PBMC abrogated PBMC adhesion to activated THBMEC. The anti-alpha4 integrin antibody, natalizumab, infused into MS patients, significantly reduced the adhesion of their ex vivo PBMC to activated THBMEC under flow conditions. Further study showed that alternatively spliced fibronectin containing the CS1 region (FN-CS1), but not Vascular Cell Adhesion Molecule type 1 (VCAM-1), was the ligand of alpha4 integrin on activated THBMEC. Blocking FN-CS1 abrogated PBMC adhesion on activated THBMEC, while anti-VCAM-1 antibodies had no effect. These results established a novel in vitro dynamic BBB model. We also demonstrated the dependence of leukocyte-endothelial interactions in this model on alpha4 integrins and FN-CS1.