Merck
GRSF1 deficiency in skeletal muscle reduces endurance in aged mice.
Aging (2021-06-04)
Riley K Driscoll, Linda K Krasniewski, Samuel G Cockey, Jen-Hao Yang, Yulan Piao, Elin Lehrmann, Yongqing Zhang, Marc Michel, Ji Heon Noh, Chang-Yi Cui, Myriam Gorospe
ABSTRACT
GRSF1 is a mitochondrial RNA-binding protein important for maintaining mitochondrial function. We found that GRSF1 is highly expressed in cultured skeletal myoblasts differentiating into myotubes. To understand the physiological function of GRSF1 in vivo, we generated mice in which GRSF1 was specifically ablated in skeletal muscle. The conditional knockout mice (Grsf1cKO) appeared normal until 7-9 months of age. Importantly, however, a reduction of muscle endurance compared to wild-type controls was observed in 16- to 18-month old Grsf1cKO mice. Transcriptomic analysis revealed more than 200 mRNAs differentially expressed in Grsf1cKO muscle at this age. Notably, mRNAs encoding proteins involved in mitochondrial function, inflammation, and ion transport, including Mgarp, Cxcl10, Nfkb2, and Sln mRNAs, were significantly elevated in aged Grsf1cKO muscle. Our findings suggest that GRSF1 deficiency exacerbates the functional decline of aged skeletal muscle, likely through multiple downstream effector proteins.
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%