Merck
  • Relationship between pectoralis major muscle histology and quality traits of chicken meat.
Relationship between pectoralis major muscle histology and quality traits of chicken meat.
Poultry science (2015-01-13)
M Mazzoni, M Petracci, A Meluzzi, C Cavani, P Clavenzani, F Sirri
ABSTRACT
A trial was conducted to evaluate the influence of myodegeneration of pectoralis major muscle on quality traits and chemical composition of breast meat of heavy-size male broilers. For this purpose, a total of 72 pectoralis major muscles were randomly collected from broilers farmed under homogeneous conditions and graded into three categories (mild, n=22; moderate, n=33; and severe, n=17) based on the presence of abnormal fibers (giant fibers, fibers with hyaline degeneration, and damaged and/or necrotic fibers) evaluated by histological and immunohistochemical analysis. Color, pH, drip loss, Allo-Kramer shear values, and chemical composition (moisture, proteins, total lipids, ashes, and collagen) were determined on nonmarinated breast meat. Purge loss and cook loss, total yield, and Allo-Kramer shear values were measured on vacuum-tumbled samples. Samples showing moderate myodegeneration had the highest mean cross-sectional area of the fibers, while samples with severe myodegeneration had myofibers of different diameter and without the characteristic polygonal shape, multifocal degeneration and necrosis, as well as infiltration of CD3-immunoreactive cells. Cooking losses of nonmarinated meat were lower in the mild group with respect to moderate and severe groups (21.4 vs. 24.7 and 24.7%; P<0.001). Breast muscles with severe damage, in comparison with mild degenerated samples, showed higher moisture (75.4 vs. 74.4%; P<0.05) and lower protein percentages (21.1 vs. 22.6%; P<0.001). The lipid percentage of severely degenerated samples was higher than that from moderate group (2.94 vs. 2.36; P<0.05), while collagen content was not modified by histological lesion levels. Marinated meat from the mild group had higher uptake and total marinade yield after cooking. In conclusion, almost all breast fillets of heavy broiler chickens produced under intensive farming systems had histological lesions, which reflected on the chemical composition of the meat and the impaired water holding/binding capacities of the meat.