- Pyrano[2,3-e]isoindol-2-ones, new angelicin heteroanalogues.
Pyrano[2,3-e]isoindol-2-ones, new angelicin heteroanalogues.
Bioorganic & medicinal chemistry letters (2009-02-24)
Paola Barraja, Virginia Spanò, Diana Patrizia, Anna Carbone, Girolamo Cirrincione, Daniela Vedaldi, Alessia Salvador, Giampietro Viola, Francesco Dall'acqua
PMID19230658
ABSTRACT
A convenient synthesis of the pyrano[2,3-e]isoindol-2-one ring system, an heteroanalogue of angelicin, is reported. Our synthetic approach consists of the annelation of the pyran ring on the isoindole moiety using 5-dialkylamino- or 5-hydroxymethylene intermediates as building blocks. The photoantiproliferative activity of the new derivatives was studied. Some of them bearing the benzyl group at the 8 position were active with IC(50) in the micromolar range. Cell cytotoxicity involves apoptosis, alteration of cell cycle profile and membrane photodamage.