Merck
  • Substituted quinazolines, part 3. Synthesis, in vitro antitumor activity and molecular modeling study of certain 2-thieno-4(3H)-quinazolinone analogs.
Substituted quinazolines, part 3. Synthesis, in vitro antitumor activity and molecular modeling study of certain 2-thieno-4(3H)-quinazolinone analogs.
European journal of medicinal chemistry (2008-10-28)
Abdulrahman M Al-Obaid, Sami G Abdel-Hamide, Hassan A El-Kashef, Alaa A-M Abdel-Aziz, Adel S El-Azab, Hamad A Al-Khamees, Hussein I El-Subbagh
ABSTRACT
The synthesis of some new 2-thieno-4(3H)-quinazolinone derivatives and their biological evaluation as antitumor agents using the National Cancer Institute (NCI) disease oriented antitumor screen protocol are investigated. Compounds 2-(2-thienylcarbonylamino)-5-iodo-N-(4-hydroxyphenyl)-benzamide (16), 2-(2-thieno)-6-iodo-3-phenylamino-3,4-dihydro-quina-zolin-4-one (26), and 2-(2-thieno)-4-[4-sulfonamidobenzylamino]-6-iodo-quinazoline (42), with GI(50) values of 12.7, 10.3, 16.9 microM, respectively, proved to be the most active members in this study, as compared to the known drug 5-FU. Conformational analysis of the most active molecules using molecular modeling and QSAR techniques enabled the understanding of the pharmacophoric requirements for 2-thieno-quinzolinone derivatives as antitumor agents. These three quinazolinone analogs (16, 26, 42) could be considered as useful templates for future development to obtain more potent antitumor agents.
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder