Merck
  • Discovery of N-{1-[3-(3-oxo-2,3-dihydrobenzo[1,4]oxazin-4-yl)propyl]piperidin-4-yl}-2-phenylacetamide (Lu AE51090): an allosteric muscarinic M1 receptor agonist with unprecedented selectivity and procognitive potential.
Discovery of N-{1-[3-(3-oxo-2,3-dihydrobenzo[1,4]oxazin-4-yl)propyl]piperidin-4-yl}-2-phenylacetamide (Lu AE51090): an allosteric muscarinic M1 receptor agonist with unprecedented selectivity and procognitive potential.
Journal of medicinal chemistry (2010-08-06)
Anette G Sams, Morten Hentzer, Gitte K Mikkelsen, Krestian Larsen, Christoffer Bundgaard, Niels Plath, Claus T Christoffersen, Benny Bang-Andersen
ABSTRACT
The discovery and structure-activity relationship (SAR) of a series of allosteric muscarinic M(1) receptor agonists are described. Compound 17 (Lu AE51090) was identified as a representative compound from the series, based on its high selectivity as an agonist at the muscarinic M(1) receptor across a panel of muscarinic receptor subtypes. Furthermore, 17 displayed a high degree of selectivity when tested in a broad panel of G-protein-coupled receptors, ion channels, transporters, and enzymes, and 17 showed an acceptable pharmacokinetic profile and sufficient brain exposure in rodents in order to characterize the compound in vivo. Hence, in a rodent model of learning and memory, 17 reversed delay-induced natural forgetting, suggesting a procognitive potential of 17.