Merck
  • Fatty acid synthase inhibitors of phenolic constituents isolated from Garcinia mangostana.
Fatty acid synthase inhibitors of phenolic constituents isolated from Garcinia mangostana.
Bioorganic & medicinal chemistry letters (2010-09-08)
He Zhong Jiang, Xiao Fang Quan, Wei Xi Tian, Jiang Miao Hu, Peng Cheng Wang, Sheng Zhuo Huang, Zhong Quan Cheng, Wen Juan Liang, Jun Zhou, Xiao Feng Ma, You Xing Zhao
ABSTRACT
Natural inhibitors of fatty acid synthase (FAS) are emerging as potential therapeutic agents to treat cancer and obesity. The bioassay-guided chemical investigation of the hulls of Garcinia mangostana led to the isolation of 13 phenolic compounds (1-13) mainly including xanthone and benzophenone, in which compounds 7, 8, 9, 10, and 11 were isolated from this plant for the first time and compound 9 was a new natural product. These isolates possess strong inhibitory activity of FAS with the IC(50) values ranging from 1.24 to 91.07 μM. The study indicates that two types of natural products, xanthones and benzophenones, could be considered as promising FAS inhibitors.
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%