Merck
  • B7-H4 induces epithelial-mesenchymal transition and promotes colorectal cancer stemness.
B7-H4 induces epithelial-mesenchymal transition and promotes colorectal cancer stemness.
Pathology, research and practice (2021-01-12)
Ying Feng, Zhaoting Yang, Chengye Zhang, Nan Che, Xingzhe Liu, Yanhua Xuan
ABSTRACT
B7-H4 is a unique negative regulator of T cells that is typically significantly overexpressed in various carcinomas and is associated with poor prognosis. However, the effects of B7-H4 expression on epithelial-mesenchymal transition (EMT) and cancer stemness of colorectal cancer (CRC) are not entirely clear. In the present study, we used tissue samples from 98 patients with CRC and CRC cell lines to determine the clinicopathological significance of B7-H4 in CRC and its effects on CRC stemness. We performed immunohistochemical staining; immunofluorescence imaging; western blotting; and tumor sphere formation, wound healing, transwell migration, and in vivo tumorigenesis assays. B7-H4 expression was upregulated in CRC tissues and was associated with lymph node metastasis, distant metastasis, clinical stage, a shorter overall survival rate, and disease-free survival rate. Cox regression analyses indicated that B7-H4 is an independent poor prognostic factor for CRC. In addition, B7-H4 expression was correlated with the expression of EMT-related proteins and cancer stemness-related proteins. Moreover, immunohistochemical and immunofluorescence analyses revealed that B7-H4 was correlated with CD133 and CD44 expression levels in both CRC tissues and HT29 and HCT116 cell lines. Conversely, B7-H4 knockdown downregulated the expression of EMT- and cancer stemness-related proteins, while inhibiting tumor spheroid formation, cell migration, and invasion of CRC cell lines. These results indicate that B7-H4 can promote EMT and may be a novel stem cell marker, suggesting its potential as a prognostic biomarker for CRC.