Merck
Pyridazinopsoralens of wide chemotherapeutic interest.
Bioorganic & medicinal chemistry (2010-07-10)
Lisa Dalla Via, Ornella Gia, Sebastiano Marciani Magno, Alessandra Braga, José Carlos González-Gómez, Lázaro Guillermo Pérez-Montoto, Eugenio Uriarte
ABSTRACT
The synthesis of new 6,10-dimethylpyridazino[4,5-h]psoralens, carrying no (4), one (5), or two (6-9) dialkylaminoalkylcarboxamide side chains on the pyridazine ring is reported. All compounds exert a significant photoantiproliferative activity. Moreover, the derivatives characterised by the protonable side chains show a notable cytotoxicity in the dark. The investigation on the mechanism of action demonstrated the capacity to intercalate into DNA base pairs and to inhibit the relaxation activity of topoisomerase II.