Merck
  • NEAT1 Knockdown Inhibits Keloid Fibroblast Progression by miR-196b-5p/FGF2 Axis.
NEAT1 Knockdown Inhibits Keloid Fibroblast Progression by miR-196b-5p/FGF2 Axis.
The Journal of surgical research (2020-11-10)
Jingzhe Yang, Pingyang Deng, Yonggang Qi, Xinshu Feng, Hailing Wen, Fengping Chen
ABSTRACT
Keloid is a benign fibroproliferative tumor of the skin caused by abnormal wound healing process after skin injury. Long noncoding RNAs have been reported to be involved in the development of keloid. However, the role and mechanism of nuclear enriched abundant transcript 1 (NEAT1) in keloid are still unknown. Quantitative real-time polymerase chain reaction was performed to detect the expression of NEAT1, miR-196b-5p, and fibroblast growth factor 2 (FGF2). Western blot was conducted to measure the levels of collagen I, α-smooth muscle actin, fibronectin, and FGF2. Cell Counting Kit-8 assay and transwell assay were used to evaluate cell viability and migration, respectively. Dual-luciferase reporter assay was conducted to verify the targeting relationship between miR-196b-5p and NEAT1 or FGF2. NEAT1 was increased and miR-196b-5p was decreased in keloid tissues and fibroblasts. NEAT1 knockdown or miR-196b-5p overexpression suppressed cell viability, migration, and extracellular matrix (ECM) component production in keloid fibroblasts. MiR-196 b-5p was a target of NEAT1, and NEAT1 overexpression reversed the effect of miR-196b-5p on keloid fibroblast progression. Moreover, we found that miR-196b-5p directly targeted FGF2. FGF2 knockdown suppressed keloid fibroblast viability, migration, and ECM protein production. FGF2 overexpression abolished the effect of miR-196b-5p overexpression on keloid fibroblast development. NEAT1 silencing suppressed cell viability, migration, and ECM expression in keloid fibroblasts by regulating miR-196b-5p/FGF2 axis, indicating a promising strategy for keloid treatment.