Merck
  • IGF1R signaling drives antiestrogen resistance through PAK2/PIX activation in luminal breast cancer.
IGF1R signaling drives antiestrogen resistance through PAK2/PIX activation in luminal breast cancer.
Oncogene (2018-01-23)
Yinghui Zhang, Lynn Wester, Jichao He, Tamar Geiger, Marja Moerkens, Ram Siddappa, Jean A Helmijr, Mieke M Timmermans, Maxime P Look, Caroline H M van Deurzen, John W M Martens, Chantal Pont, Marjo de Graauw, Erik H J Danen, Els M J J Berns, John H N Meerman, Maurice P H M Jansen, Bob van de Water
ABSTRACT
Antiestrogen resistance in estrogen receptor positive (ER+) breast cancer is associated with increased expression and activity of insulin-like growth factor 1 receptor (IGF1R). Here, a kinome siRNA screen has identified 10 regulators of IGF1R-mediated antiestrogen with clinical significance. These include the tamoxifen resistance suppressors BMPR1B, CDK10, CDK5, EIF2AK1, and MAP2K5, and the tamoxifen resistance inducers CHEK1, PAK2, RPS6KC1, TTK, and TXK. The p21-activated kinase 2, PAK2, is the strongest resistance inducer. Silencing of the tamoxifen resistance inducing genes, particularly PAK2, attenuates IGF1R-mediated resistance to tamoxifen and fulvestrant. High expression of PAK2 in ER+ metastatic breast cancer patients is correlated with unfavorable outcome after first-line tamoxifen monotherapy. Phospho-proteomics has defined PAK2 and the PAK-interacting exchange factors PIXα/β as downstream targets of IGF1R signaling, which are independent from PI3K/ATK and MAPK/ERK pathways. PAK2 and PIXα/β modulate IGF1R signaling-driven cell scattering. Targeting PIXα/β entirely mimics the effect of PAK2 silencing on antiestrogen re-sensitization. These data indicate PAK2/PIX as an effector pathway in IGF1R-mediated antiestrogen resistance.