- Distribution of ozone and related compounds in the marine boundary layer of the Northern South China Sea in 2010
Distribution of ozone and related compounds in the marine boundary layer of the Northern South China Sea in 2010
Aerosol and Air Quality Research (2015)
Lan YY, et al.