Merck
  • Possible participation of superoxide anion in the intestinal tryptophan 2,3-dioxygenase reaction.
Possible participation of superoxide anion in the intestinal tryptophan 2,3-dioxygenase reaction.
The Journal of biological chemistry (1971-12-25)
F Hirata, O Hayaishi