Merck
  • Bovine erythrocyte superoxide dismutase: diazo coupling, subunit interactions, and electrophoretic variants.
Bovine erythrocyte superoxide dismutase: diazo coupling, subunit interactions, and electrophoretic variants.
Biochemistry (1979-01-09)
D P Malinowski, I Fridovich
PMID420775