Merck
  • Maternal conjugated linoleic acid consumption prevented TAG alterations induced by a high-fat diet in male adult rat offspring.
Maternal conjugated linoleic acid consumption prevented TAG alterations induced by a high-fat diet in male adult rat offspring.
The British journal of nutrition (2020-04-03)
Marcela Aída González, Jimena Lavandera, Carolina Gerstner, Ana Clara Fariña, Juliana Saín, Claudio Adrián Bernal
ABSTRACT
Maternal nutritional programming by a high-fat (HF) diet is related to hepatic lipid accumulation and steatosis in offspring. Conjugated linoleic acid (CLA) might ameliorate impaired hepatic lipid homoeostasis; therefore, the aim was to investigate the potential preventive effect of maternal CLA consumption on TAG metabolism alterations induced by HF diets in adult male rat offspring receiving or not receiving CLA. Female Wistar rats were fed a control (C) diet, HF diet or HF diet supplemented with CLA (HF+CLA) for 4 weeks before mating and throughout pregnancy and lactation. After weaning, for 9 weeks, male offspring of C or HF rats continued with the same diets as their mothers (C/C or HF/HF groups, respectively) and male offspring of HF+CLA rats were fed HF or HF+CLA diets (HF+CLA/HF or HF+CLA/HF+CLA groups, respectively). Nutritional parameters, serum and liver TAG levels, the TAG secretion rate (TAG-SR) and the activities as well as gene expression of key hepatic enzymes involved in TAG regulation were assessed. The most interesting results were that maternal CLA decreased epididymal white adipose tissue weight and prevented serum and liver TAG accumulation induced by a HF diet in adult male offspring receiving or not receiving CLA. The prevention of liver steatosis in HF+CLA/HF+CLA and HF+CLA/HF offspring was associated with an increased hepatic TAG-SR. Overall, this study provides evidence that maternal CLA consumption programmes TAG regulation and in this way contributes to lowering lipid levels in tissues and preventing liver steatosis in particular.
Sigma-Aldrich
Anti-Cdc25C Antibody, clone TC-15, clone TC-15, Upstate®, from mouse