Merck
  • Identification and characterization of autosomal genes that interact with glass in the developing Drosophila eye.
Identification and characterization of autosomal genes that interact with glass in the developing Drosophila eye.
Genetics (1996-04-01)
C Ma, H Liu, Y Zhou, K Moses
ABSTRACT
The glass gene encodes a zinc finger, DNA-binding protein that is required for photoreceptor cell development in Drosophila melanogaster. In the developing compound eye, glass function is regulated at two points: (1) the protein is expressed in all cells' nuclei posterior to the morphogenetic furrow and (2) the ability of the Glass protein to regulate downstream genes is largely limited to the developing photoreceptor cells. We conducted a series of genetic screens for autosomal dominant second-site modifiers of the weal allele glass3, to discover genes with products that may regulate glass function at either of these levels. Seventy-six dominant enhancer mutations were recovered (and no dominant suppressors). Most of these dominant mutations are in essential genes and are associated with recessive lethality. We have assigned these mutations to 23 complementation groups that include multiple alleles of Star and hedgehog as well as single alleles of Delta, roughened eye, glass and hairy. Mutations in 18 of the complementation groups are embryonic lethals, and of these, 13 show abnormal adult retinal phenotypes in homozygous clones, usually with altered numbers of photoreceptor cells in some of the ommatidia.