Merck
  • Non-invasive Protocol for Kinematic Monitoring of Root Growth under Infrared Light.
Non-invasive Protocol for Kinematic Monitoring of Root Growth under Infrared Light.
Bio-protocol (2017-07-20)
François Bizet, Lionel X Dupuy, Anthony Glyn Bengough, Alexis Peaucelle, Irène Hummel, Marie-Béatrice Bogeat-Triboulot
ABSTRACT
Phenotyping the dynamics of root responses to environmental cues is necessary to understand plant acclimation to their environment. Continuous monitoring of root growth is challenging because roots normally grow belowground and are very sensitive to their growth environment. This protocol combines infrared imaging with hydroponic cultivation for kinematic analyses. It allows continuous imaging at fine spatiotemporal resolution and disturbs roots minimally. Examples are provided of how the procedure and materials can be adapted for 3D monitoring and of how environmental stress may be manipulated for experimental purposes.