Merck
  • Assessing the Optimum Level of Supplementation with Camelina Seeds in Ewes' Diets to Improve Milk Quality.
Assessing the Optimum Level of Supplementation with Camelina Seeds in Ewes' Diets to Improve Milk Quality.
Foods (Basel, Switzerland) (2021-09-29)
Christos Christodoulou, Alexandros Mavrommatis, Christina Mitsiopoulou, George Symeon, Vasilis Dotas, Kyriaki Sotirakoglou, Basiliki Kotsampasi, Eleni Tsiplakou
ABSTRACT
Camelina sativa seeds are rich in bioactive compounds such as polyunsaturated fatty acids (PUFA) and antioxidants, thus, their supplementation in ewes' diets, may be an effective way to develop high nutritional dairy products. Therefore, the present study investigates the effect of the dietary inclusion of Camelina sativa seeds in ewes' oxidative status and milk quality. Forty-eight dairy Chios ewes were divided into four homogenous groups and were fed individually. The concentrate of the control group (CON) had no inclusion of Camelina seeds, while the treatment groups (CSS6, CSS11, CSS16) were supplemented with 6%, 11%, and 16%, respectively. Including Camelina seeds in 6% and 11%, had no impact on milk performance, while in the CSS16, milk fat was significantly decreased compared to the CON. Supplementing Camelina seeds improved milk quality from a human health perspective by modifying the content of saturated fatty acid, the proportions of α-linolenic (C18:3 n-3), and C18:2 cis-9, trans-11 (CLA), and the ω6/ω3 ratio. Furthermore, the activity of catalase (CAT) was significantly increased in the CSS11 and CSS16, and superoxide dismutase (SOD) activity also significantly upsurged in the CSS16. Still, the levels of malondialdehyde (MDA) were significantly increased in the CSS11 compared to the CON and CSS6, and in the CSS16 compared to the CSS6. In CSS16, protein carbonyls were significantly increased. Finally, in the CSS-fed ewes, milk oxidative stability was fortified, as suggested by the modifications in the activities of SOD, CAT, and glutathione peroxidase (GSH-Px), in the antioxidant capacity, and the oxidative stress biomarkers. Consequently, the incorporation of 6% Camelina seeds in the concentrates of ewes improves milk's fatty acid profile and oxidative status. However, more research is required regarding the possible negative effects of the constant consumption of Camelina seeds by ewes.