Merck
  • Identification of novel proteins and mechanistic pathways associated with early-onset hypertension by deep proteomic mapping of resistance arteries.
Identification of novel proteins and mechanistic pathways associated with early-onset hypertension by deep proteomic mapping of resistance arteries.
The Journal of biological chemistry (2021-12-21)
Joakim A Bastrup, Christian Aalkjær, Thomas A Jepps
ABSTRACT
Resistance arteries are small blood vessels that create resistance to blood flow. In hypertension, resistance arteries undergo remodeling, affecting their ability to contract and relax appropriately. To date, no study has mapped the hypertension-related proteomic changes in resistance arteries. Using a novel data-independent acquisition-mass spectrometry (DIA-MS) approach, we determined the proteomic changes in small mesenteric and renal arteries in pre- and early-onset hypertension from the spontaneously hypertensive rat (SHR) model, which represents human primary hypertension. Compared with normotensive controls, mesenteric arteries from 12-week-old SHRs had 286 proteins that were significantly up- or downregulated, whereas 52 proteins were identified as up- or downregulated in mesenteric arteries from 6-week-old SHRs. Of these proteins, 18 were also similarly regulated in SHR renal arteries. Our pathway analyses reveal several novel pathways in the pathogenesis of hypertension. Finally, using a matrisome database, we identified 38 altered extracellular-matrix-associated proteins, many of which have never previously been associated with hypertension. Taken together, this study reveals novel proteins and mechanisms that are associated with early-onset hypertension, thereby providing novel insights into disease progression.