Merck
Change of saliva composition with radiotherapy.
Archives of oral biology (2019-07-22)
Vera J Müller, Georgios N Belibasakis, Philipp P Bosshard, Daniel B Wiedemeier, Dominique Bichsel, Martin Rücker, Bernd Stadlinger
ABSTRACT
The aim of this study was to analyze the physiological and microbiological changes of saliva from patients with head and neck cancer during and after intensity-modulated radiotherapy (IMRT). In this prospective clinical trial saliva samples and oral candida swabs were collected from patients receiving IMRT due to head and neck cancer (examination group). The first measurement was scheduled before radiotherapy, the other measurements during and after radiotherapy up to a one year follow-up. Additionally samples from healthy controls were collected over six weeks. Salivary flow rate and pH were measured. Microbiological analysis of cariogenic and periodontopathogenic taxa was performed by fluorescence in situ hybridization and oral Candida spp occurrence was evaluated by swab tests. 11 patients and 19 controls were included. The salivary flow rate and the unstimulated pH of the examination group were significantly reduced during radiotherapy compared with the measurement before radiotherapy and to the control group. Total bacteria, streptococci and lactobacilli numbers slightly increased after radiotherapy, resuming baseline levels after one year. Mutans streptococci, Porphyromonas gingivalis and Treponema denticola were barely detectable, whereas Tannerella forsythia slightly increased following radiotherapy. No differences in Candida levels were observed in the study. Salivary changes in quantitative, qualitative and microbial composition occur during and after radiotherapy, with resumption of the measurements towards baseline levels after one year. While low levels of cariogenic and periodontopathogenic species were detected, the lower pH and salivary flow combined with increased numbers of aciduric and acidogenic lactobacilli corroborates a higher risk for caries, necessitating prevention.