Merck
  • Immunogenicity and Safety of SARS-CoV-2 mRNA Vaccines in a Cohort of Patients With Type 1 Diabetes.
Immunogenicity and Safety of SARS-CoV-2 mRNA Vaccines in a Cohort of Patients With Type 1 Diabetes.
Diabetes (2022-05-14)
Francesca D'Addio, Gianmarco Sabiu, Vera Usuelli, Emma Assi, Ahmed Abdelsalam, Anna Maestroni, Andy Joe Seelam, Moufida Ben Nasr, Cristian Loretelli, Davide Mileto, Giada Rossi, Ida Pastore, Laura Montefusco, Paola S Morpurgo, Laura Plebani, Antonio Rossi, Enrica Chebat, Andrea M Bolla, Maria Elena Lunati, Chiara Mameli, Maddalena Macedoni, Spinello Antinori, Stefano Rusconi, Maurizio Gallieni, Cesare Berra, Franco Folli, Massimo Galli, Maria Rita Gismondo, Gianvincenzo Zuccotti, Paolo Fiorina
ABSTRACT
Patients with type 1 diabetes (T1D) may develop severe outcomes during coronavirus disease 2019 (COVID-19), but their ability to generate an immune response against the SARS-CoV-2 mRNA vaccines remains to be established. We evaluated the safety, immunogenicity, and glycometabolic effects of the severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) mRNA vaccines in patients with T1D. A total of 375 patients (326 with T1D and 49 subjects without diabetes) who received two doses of the SARS-CoV-2 mRNA vaccines (mRNA-1273, BNT162b2) between March and April 2021 at ASST Fatebenefratelli Sacco were included in this monocentric observational study. Local and systemic adverse events were reported in both groups after SARS-CoV-2 mRNA vaccination, without statistical differences between them. While both patients with T1D and subjects without diabetes exhibited a parallel increase in anti-SARS-CoV-2 spike titers after vaccination, the majority of patients with T1D (70% and 78%, respectively) did not show any increase in the SARS-CoV-2-specific cytotoxic response compared with the robust increase observed in all subjects without diabetes. A reduced secretion of the T-cell-related cytokines interleukin-2 and tumor necrosis factor-α in vaccinated patients with T1D was also observed. No glycometabolic alterations were evident in patients with T1D using continuous glucose monitoring during follow-up. Administration of the SARS-CoV-2 mRNA vaccine is associated with an impaired cellular SARS-CoV-2-specific cytotoxic immune response in patients with T1D.