Merck
  • AMPK-related protein kinase ARK5 regulates subcellular localization of RNA-binding protein hnRNP A1 during hypertonic stress.
AMPK-related protein kinase ARK5 regulates subcellular localization of RNA-binding protein hnRNP A1 during hypertonic stress.
The Journal of biological chemistry (2022-08-14)
Krishna Bhattarai, Travis Richard, Thet Fatica, Brianna Frangione, William G Willmore, Martin Holcik
ABSTRACT
The heterogeneous nuclear ribonucleoprotein hnRNP A1 is a nucleocytoplasmic-shuttling RNA-binding protein that plays an important role in nucleic acid metabolism and gene expression regulation. The function of hnRNP A1 is determined in part by its specific location within the cell. Although some work has been done to elucidate the signaling pathways that regulate the cellular localization of hnRNP A1, the precise mechanism(s), including physiological and pathophysiological conditions that alter hnRNP A1 localization, are not known. We previously conducted an unbiased RNAi-based kinome-wide screen to identify kinases that regulate hnRNP A1 localization during hypertonic stress. One of the hits from this screen is AMPK-related protein kinase 5 (ARK5). Here, we validate ARK5 as the kinase responsible for controlling hnRNP A1 subcellular localization in response to hypertonic stress. We find using immunoprecipitation and in vitro kinase assay methods that ARK5 directly interacts with and phosphorylates hnRNP A1 on serine residues within the F-peptide region. We further show that the M9 motif of hnRNP A1 is essential for the ARK5-hnRNP A1 interaction and subsequent phosphorylation. In addition, the silencing of ARK5 increases the expression of antiapoptotic protein Bcl-xL and consequently delays caspase activation during hypertonic stress. Our results indicate that ARK5 phosphorylates hnRNP A1 and regulates its subcellular localization during hypertonic stress.