Merck
  • EGFR-mediated activation of adipose tissue macrophages promotes obesity and insulin resistance.
EGFR-mediated activation of adipose tissue macrophages promotes obesity and insulin resistance.
Nature communications (2022-08-11)
Shirong Cao, Yu Pan, Jiaqi Tang, Andrew S Terker, Juan Pablo Arroyo Ornelas, Guan-Nan Jin, Yinqiu Wang, Aolei Niu, Xiaofeng Fan, Suwan Wang, Raymond C Harris, Ming-Zhi Zhang
ABSTRACT
Obesity and obesity-related health complications are increasing in prevalence. Adipose tissue from obese subjects has low-grade, chronic inflammation, leading to insulin resistance. Adipose tissue macrophages (ATMs) are a source of proinflammatory cytokines that further aggravate adipocyte dysfunction. In response to a high fat diet (HFD), ATM numbers initially increase by proliferation of resident macrophages, but subsequent increases also result from infiltration in response to chemotactic signals from inflamed adipose tissue. To elucidate the underlying mechanisms regulating the increases in ATMs and their proinflammatory phenotype, we investigated the role of activation of ATM epidermal growth factor receptor (EGFR). A high fat diet increased expression of EGFR and its ligand amphiregulin in ATMs. Selective deletion of EGFR in ATMs inhibited both resident ATM proliferation and monocyte infiltration into adipose tissue and decreased obesity and development of insulin resistance. Therefore, ATM EGFR activation plays an important role in adipose tissue dysfunction.
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies