Merck
Targeting firing rate neuronal homeostasis can prevent seizures.
Disease models & mechanisms (2022-09-09)
Fred Mulroe, Wei-Hsiang Lin, Connie Mackenzie-Gray Scott, Najat Aourz, Yuen Ngan Fan, Graham Coutts, R Ryley Parrish, Ilse Smolders, Andrew Trevelyan, Robert C Wykes, Stuart Allan, Sally Freeman, Richard A Baines
ABSTRACT
Manipulating firing-rate neuronal homeostasis, which enables neurons to regulate their intrinsic excitability, offers an attractive opportunity to prevent seizures. However, to date, no drug-based interventions have been reported that manipulate this type of neuronal homeostatic mechanism. Here, we used a combination of Drosophila and mouse, and, in the latter, both a pentylenetetrazole (PTZ)-induced seizure model and an electrically induced seizure model for refractory seizures to evaluate the anticonvulsant efficacy of a novel class of anticonvulsant compounds, based on 4-tert-butyl-benzaldehyde (4-TBB). The mode of action included increased expression of the firing rate homeostatic regulator Pumilio (PUM). Knockdown of pum expression, in Drosophila, blocked anticonvulsive effects of 4-TBB, while analysis of validated PUM targets in mouse brain revealed significant reductions following exposure to this compound. A structure-activity study identified the active parts of the molecule and, further, showed that the pyrazole analogue demonstrates highest efficacy, being active against both PTZ-induced and electrically induced seizures. This study provides a proof of principle that anticonvulsant effects can be achieved through regulation of firing rate neuronal homeostasis and identifies a possible chemical compound for future development.