Merck
  • Trappc1 deficiency impairs thymic epithelial cell development by breaking endoplasmic reticulum homeostasis.
Trappc1 deficiency impairs thymic epithelial cell development by breaking endoplasmic reticulum homeostasis.
European journal of immunology (2022-08-01)
Xue Dong, Zhanfeng Liang, Jiayu Zhang, Qian Zhang, Yanan Xu, Zhaoqi Zhang, Lianfeng Zhang, Baojun Zhang, Yong Zhao
ABSTRACT
Thymic epithelial cells (TECs) are important for T cell development and immune tolerance establishment. Although comprehensive molecular regulation of TEC development has been studied, the role of transport protein particle complexes (Trappcs) in TECs is not clear. Using TEC-specific homozygous or heterozygous Trappc1 deleted mice model, we find that Trappc1 deficiency cause severe thymus atrophy with decreased cell number and blocked maturation of TECs. Mice with a TEC-specific Trappc1 deletion show poor thymic T cell output and have a greater percentage of activated/memory T cells, suffered from spontaneous autoimmune disorders. Our RNA-seq and molecular studies indicated that the decreased endoplasmic reticulum (ER) and Golgi apparatus, enhanced unfolded protein response (UPR) and subsequent Atf4-CHOP-mediated apoptosis, and reactive oxygen species (ROS)-mediated ferroptosis coordinately contributed to the reduction of Trappc1-deleted TECs. Additionally, reduced Aire+ mTECs accompanied by the decreased expression of Irf4, Irf8, and Tbx21 in Trappc1 deficiency mTECs, may further coordinately block the tissue-restricted antigen expression. In this study, we reveal that Trappc1 plays an indispensable role in TEC development and maturation and provide evidence for the importance of inter-organelle traffic and ER homeostasis in TEC development.