Merck
  • KIFC1 regulates ZWINT to promote tumor progression and spheroid formation in colorectal cancer.
KIFC1 regulates ZWINT to promote tumor progression and spheroid formation in colorectal cancer.
Pathology international (2021-04-06)
Shintaro Akabane, Naohide Oue, Yohei Sekino, Ryuichi Asai, Pham Quoc Thang, Daiki Taniyama, Kazuhiro Sentani, Masashi Yukawa, Takashi Toda, Ken-Ichi Kimura, Hiroyuki Egi, Wataru Shimizu, Hideki Ohdan, Wataru Yasui
ABSTRACT
Colorectal cancer (CRC) is the second leading cause of cancer-related mortality worldwide. Kinesin Family Member C1 (KIFC1) has been proposed as a promising therapeutic target due to its pivotal role in centrosome clustering to mediate cancer cell progression. This study aimed to analyze the expression and biological function of KIFC1 in CRC. Immunohistochemically, 67 (52%) of 129 CRC cases were positive for KIFC1 and statistically associated with poorer overall survival. KIFC1 small interfering RNA (siRNA)-transfected cells demonstrated lower cell proliferation as compared to the negative control cells. A specific KIFC1 inhibitor, kolavenic acid analog (KAA) drastically inhibited CRC cell proliferation. Microarray analysis revealed that KAA-treated CRC cells presented reduced ZW10 interacting kinetochore protein (ZWINT) expression as compared to control cells. Immunohistochemical analysis demonstrated that 61 (47%) of 129 CRC cases were positive for ZWINT and ZWINT expression was significantly correlated with KIFC1 expression. ZWINT-positive cases exhibited significantly worse overall survival. KIFC1 siRNA-transfected cells showed reduced ZWINT expression while ZWINT siRNA-transfected cells decreased cell proliferation. Both KIFC1 and ZWINT knockdown cells attenuated spheroid formation ability. This study provides new insights into KIFC1 regulating ZWINT in CRC progression and its potential as a therapeutic target.