Merck
  • Molecular basis of Mg2+ permeation through the human mitochondrial Mrs2 channel.
Molecular basis of Mg2+ permeation through the human mitochondrial Mrs2 channel.
Nature communications (2023-08-06)
Ming Li, Yang Li, Yue Lu, Jianhui Li, Xuhang Lu, Yue Ren, Tianlei Wen, Yaojie Wang, Shenghai Chang, Xing Zhang, Xue Yang, Yuequan Shen
ABSTRACT
Mitochondrial RNA splicing 2 (Mrs2), a eukaryotic CorA ortholog, enables Mg2+ to permeate the inner mitochondrial membrane and plays an important role in mitochondrial metabolic function. However, the mechanism by which Mrs2 permeates Mg2+ remains unclear. Here, we report four cryo-electron microscopy (cryo-EM) reconstructions of Homo sapiens Mrs2 (hMrs2) under various conditions. All of these hMrs2 structures form symmetrical pentamers with very similar pentamer and protomer conformations. A special structural feature of Cl--bound R-ring, which consists of five Arg332 residues, was found in the hMrs2 structure. Molecular dynamics simulations and mitochondrial Mg2+ uptake assays show that the R-ring may function as a charge repulsion barrier, and Cl- may function as a ferry to jointly gate Mg2+ permeation in hMrs2. In addition, the membrane potential is likely to be the driving force for Mg2+ permeation. Our results provide insights into the channel assembly and Mg2+ permeation of hMrs2.