Merck
  • Novel biallelic mutations in TTC29 cause asthenoteratospermia and male infertility.
Novel biallelic mutations in TTC29 cause asthenoteratospermia and male infertility.
Molecular genetics & genomic medicine (2022-11-09)
Siyu Dai, Yan Liang, Mohan Liu, Yanting Yang, Hongqian Liu, Ying Shen
ABSTRACT
Multiple morphological abnormalities of the sperm flagella (MMAF), which is characterized as asthenoteratospermia involving absent, short, bent, coiled, and/or irregular-caliber flagella, is a rare recessive inherited disorder associated with male infertility. To date, genetic causes of MMAF cases are not fully explored. Whole-exome sequencing was conducted to identify pathogenic variants in a patient with MMAF. The functional effect of the identified mutations was investigated by immunofluorescence staining and western blotting. Intracytoplasmic sperm injection was used to assist fertilization for the patient with MMAF. We identified novel biallelic mutations, a splicing variant NC_000004.12:g.146937593C>T (c.254+1G>A), and a nonsense mutation NM_001300761.4:c.1185C>G (NP_001287690.1:p.Tyr395*), in TTC29 from an infertile patient. In addition to the typical MMAF phenotype, the patient also presented aberrant morphology of sperm heads. Further functional experiments confirmed the absence of TTC29 expression in the spermatozoa. We also explored the specific expression pattern of TTC29 in human and mouse spermatogenesis. The outcome of intracytoplasmic sperm injection in the patient was unsuccessful, while additional female risk factors should not be excluded. Our study revealed the novel biallelic mutations in TTC29 in a MMAF patient, which findings expand the mutational spectrum of TTC29 and further contribute to the diagnosis, genetic counseling, and prognosis of male infertility.