Merck
  • Protocol for combining immunological procedures with cell cycle stage synchronization to detect cell-cycle-specific complexes.
Protocol for combining immunological procedures with cell cycle stage synchronization to detect cell-cycle-specific complexes.
STAR protocols (2023-03-01)
Toshiyuki Habu, Jiyeong Kim
ABSTRACT
Here, we present a protocol combining co-immunoprecipitation (Co-IP) and immunofluorescence approaches with cell cycle stage synchronization to detect cell-cycle-specific complexes. We describe steps to synchronize cells at specific cell cycle stages using drugs. We then detail the preparation of cell extracts from synchronized cells and fractionation of the protein complexes with density centrifugation, followed by Co-IP with specific antibodies. Protein-protein interactions are confirmed by localization using immunofluorescence imaging. This protocol is helpful for visualizing the dynamics of protein complex assembly. For complete details on the use and execution of this protocol, please refer to Habu and Kim (2021).1.
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)