Merck
  • TP53TG1/STAT axis is involved in the development of colon cancer through affecting PD-L1 expression and immune escape mechanism of tumor cells.
TP53TG1/STAT axis is involved in the development of colon cancer through affecting PD-L1 expression and immune escape mechanism of tumor cells.
American journal of cancer research (2023-12-07)
Wenmin Ji, Wenyan Wang, Yanfang Wei
ABSTRACT
This research is dedicated to investigating the mechanism of programmed cell death ligand 1 (PD-L1) and tumor protein 53 target gene 1 (TP53TG1) in immune regulation of colon cancer (CC). Expressions of TP53TG1, PD-L1 and signal transducers and activators of transcription (STATs) in CC and their correlation were detected through bioinformatics analysis. Effects of PD-L1 and TP53TG1 on the CC were assessed by in vivo and in vitro experiments. Herein, PD-L1 level was negatively correlated with TP53TG1 expression, but was positively correlated with the levels of STATs. Both overexpressed TP53TG1 and PD-L1 antibody reversed the effects of CT26 cells on inhibiting cell proliferation, cytokine secretion and PD-L1 level, and enhancing the cytotoxicity of NK cells and CD8+ T cells. TP53TG1 reduced PD-L1 level by inactivating STATs pathway. Downregulation of PD-L1 increased cytokine secretion and T lymphocyte killing ability, promoted tumor cell apoptosis, and inhibited the tumor growth. Altogether, TP53TG1/STAT axis regulates the immunomodulatory mechanism of CC by reducing PD-L1 expression.