Merck
  • Isolated Chronic Mucocutaneous Candidiasis due to a Novel Duplication Variant of IL17RC.
Isolated Chronic Mucocutaneous Candidiasis due to a Novel Duplication Variant of IL17RC.
Journal of clinical immunology (2023-12-22)
Kosuke Noma, Miyuki Tsumura, Tina Nguyen, Takaki Asano, Fumiaki Sakura, Moe Tamaura, Yusuke Imanaka, Yoko Mizoguchi, Shuhei Karakawa, Seiichi Hayakawa, Takayo Shoji, Junichi Hosokawa, Kazushi Izawa, Yun Ling, Jean-Laurent Casanova, Anne Puel, Stuart G Tangye, Cindy S Ma, Osamu Ohara, Satoshi Okada
ABSTRACT
Inborn errors of the IL-17A/F-responsive pathway lead to chronic mucocutaneous candidiasis (CMC) as a predominant clinical phenotype, without other significant clinical manifestations apart from mucocutaneous staphylococcal diseases. Among inborn errors affecting IL-17-dependent immunity, autosomal recessive (AR) IL-17RC deficiency is a rare disease with only three kindreds described to date. The lack of an in vitro functional evaluation system of IL17RC variants renders its diagnosis difficult. We sought to characterize a 7-year-old Japanese girl with CMC carrying a novel homozygous duplication variant of IL17RC and establish a simple in vitro system to evaluate the impact of this variant. Flow cytometry, qPCR, RNA-sequencing, and immunoblotting were conducted, and an IL17RC-knockout cell line was established for functional evaluation. The patient presented with oral and mucocutaneous candidiasis without staphylococcal diseases since the age of 3 months. Genetic analysis showed that the novel duplication variant (Chr3: 9,971,476-9,971,606 dup (+131bp)) involving exon 13 of IL17RC results in a premature stop codon (p.D457Afs*16 or p.D457Afs*17). Our functional evaluation system revealed this duplication to be loss-of-function and enabled discrimination between loss-of-function and neutral IL17RC variants. The lack of response to IL-17A by the patient's SV40-immortalized fibroblasts was restored by introducing WT-IL17RC, suggesting that the genotype identified is responsible for her clinical phenotype. The clinical and cellular phenotype of the current case of AR IL-17RC deficiency supports a previous report on this rare disorder. Our newly established evaluation system will be useful for the diagnosis of AR IL-17RC deficiency, providing accurate validation of unknown IL17RC variants.