Merck
  • Human islet amyloid polypeptide-induced β-cell cytotoxicity is linked to formation of α-sheet structure.
Human islet amyloid polypeptide-induced β-cell cytotoxicity is linked to formation of α-sheet structure.
Protein science : a publication of the Protein Society (2023-12-08)
Cheng-Chieh Hsu, Andrew T Templin, Tatum Prosswimmer, Dylan Shea, Jinzheng Li, Barbara Brooks-Worrell, Steven E Kahn, Valerie Daggett
ABSTRACT
Type 2 diabetes (T2D) results from insulin secretory dysfunction arising in part from the loss of pancreatic islet β-cells. Several factors contribute to β-cell loss, including islet amyloid formation, which is observed in over 90% of individuals with T2D. The amyloid is comprised of human islet amyloid polypeptide (hIAPP). Here we provide evidence that early in aggregation, hIAPP forms toxic oligomers prior to formation of amyloid fibrils. The toxic oligomers contain α-sheet secondary structure, a nonstandard secondary structure associated with toxic oligomers in other amyloid diseases. De novo, synthetic α-sheet compounds designed to be nontoxic and complementary to the α-sheet structure in the toxic oligomers inhibit hIAPP aggregation and neutralize oligomer-mediated cytotoxicity in cell-based assays. In vivo administration of an α-sheet design to mice for 4 weeks revealed no evidence of toxicity nor did it elicit an immune response. Furthermore, the α-sheet designs reduced endogenous islet amyloid formation and mitigation of amyloid-associated β-cell loss in cultured islets isolated from an hIAPP transgenic mouse model of islet amyloidosis. Characterization of the involvement of α-sheet in early aggregation of hIAPP and oligomer toxicity contributes to elucidation of the molecular mechanisms underlying amyloid-associated β-cell loss.