Merck
  • Comparative Analysis of Apigenin-3 Acetate versus Apigenin and Methyl-Prednisolone in Inhibiting Proliferation and Gene Expression of Th1 Cells in Multiple Sclerosis.
Comparative Analysis of Apigenin-3 Acetate versus Apigenin and Methyl-Prednisolone in Inhibiting Proliferation and Gene Expression of Th1 Cells in Multiple Sclerosis.
Cell journal (2023-06-10)
Neda Kasiri, Seyed Mostafa Ghannadian, Reza Hosseini, Leila Ahmadi, Mahshid Rahmati, Fereshteh Ashtari, Abbasali Pourazar, Nahid Eskandari
ABSTRACT
In spite of the advances in therapeutic modalities, morbidity, due to multiple sclerosis (MS), still remains high. Therefore, a large body of research is endeavouring to discover or develop novel therapies with improved efficacy for treating MS patients. In the present study, we examined the immunomodulatory effects of apigenin (Api) on peripheral blood mononuclear cells (PBMCs) isolated from MS patients. We also developed an acetylated form of Api (apigenin- 3-acetate) to improve In its blood-brain barrier (BBB) permeability. Additionally, we compared its anti-inflammatory properties to original Api and methyl-prednisolone-acetate (a standard therapy), as a potential option in treating MS patients. The current study was an experimental-interventional research. The half maximal inhibitory concentration (IC50) values for apigenin-3-acetate, apigenin, and methyl-prednisolone-acetate were determined in healthy volunteers' PBMCs (n=3). Gene expressions of T-box transcription factor (TBX21 or T-bet) and IFN-γ, as well as proliferation of T cells isolated from MS patients' PBMCs (n=5), were examined in co-cultures of apigenin-3-acetate, Api and methyl-prednisolone-acetate after 48 hours of treatment, using quantitative reverse transcription polymerase chain reaction (qRT-PCR). Our findings showed that apigenin-3-acetate, apigenin, and methyl-prednisolone-acetate at concentrations of 80, 80, and 2.5 M could inhibit Th1 cell proliferation after 48 hours (P=0.001, P=0.036, and P=0.047, respectively); they also inhibited T-bet (P=0.015, P=0.019, and P=0.022) and interferon-γ (IFN-γ) gene expressions (P=0.0001). Our findings suggested that Api may have anti-inflammatory properties, possibly by inhibiting proliferation of IFN-producing Th1 cells. Moreover, comparative immunomodulatory effects were found for the acetylated version of apigenin-3-acetate versus Api and methyl-prednisolone-acetate.