Merck
  • Palladium-catalyzed C-N cross coupling of sulfinamides and aryl halides.
Palladium-catalyzed C-N cross coupling of sulfinamides and aryl halides.
The Journal of organic chemistry (2012-03-31)
Xiaofei Sun, Xingzhao Tu, Chuan Dai, Xiaoping Zhang, Binbin Zhang, Qingle Zeng
ABSTRACT
The palladium-catalyzed C-N cross coupling of sulfinamides and aryl halides is reported. In the presence of Pd(2)(dba)(3), tBuXPhos, NaOH, and a small amount of water, the C-N cross coupling of chiral tert-butanesulfinamide and aryl halides was accomplished to give N-aryl tert-butanesulfinamide without racemization, and the coupling of racemic p-toluenesulfinamide smoothly afforded N-aryl p-toluenesulfinamides. 2-Bromopyridine was also suitable for the coupling. Addition of a small amount of water to the catalytic system was of importance to obtain high yields.