Merck
  • SGCE promotes breast cancer stemness by promoting the transcription of FGF-BP1 by Sp1.
SGCE promotes breast cancer stemness by promoting the transcription of FGF-BP1 by Sp1.
The Journal of biological chemistry (2023-10-15)
Ting Qiu, Lei Hou, Lina Zhao, Xinye Wang, Zhongmei Zhou, Chuanyu Yang, Huifeng Zhang, Dewei Jiang, Baowei Jiao, Ceshi Chen
ABSTRACT
Breast cancer stem cells are mainly responsible for poor prognosis, especially in triple-negative breast cancer (TNBC). In a previous study, we demonstrated that ε-Sarcoglycan (SGCE), a type Ⅰ single-transmembrane protein, is a potential oncogene that promotes TNBC stemness by stabilizing EGFR. Here, we further found that SGCE depletion reduces breast cancer stem cells, partially through inhibiting the transcription of FGF-BP1, a secreted oncoprotein. Mechanistically, we demonstrate that SGCE could interact with the specific protein 1 transcription factor and translocate into the nucleus, which leads to an increase in the transcription of FGF-BP1, and the secreted FBF-BP1 activates FGF-FGFR signaling to promote cancer cell stemness. The novel SGCE-Sp1-FGF-BP1 axis provides novel potential candidate diagnostic markers and therapeutic targets for TNBC.