Merck
  • Regulation of hPCL3 isoforms' ubiquitination by TRIM21 in non-small cell lung cancer progression.
Regulation of hPCL3 isoforms' ubiquitination by TRIM21 in non-small cell lung cancer progression.
Life science alliance (2023-07-29)
Ye Xu, Wenhong Liu, Xiawei Jiang, Junfeng Li, Qingling Liu, Fang Su, Shanming Ruan, Zhiqian Zhang, Fangfang Tao
ABSTRACT
Non-small cell lung cancer (NSCLC) is the main subtype of lung cancer. The role of hPCL3 isoforms, hPCL3S and hPCL3L, remains ambiguous. This study examines the functional implications of these isoforms in NSCLC, using lung cancer cell lines A549 and NCI-H226c for in vivo and in vitro analyses. The results indicate that elevated expression of both hPCL3S and hPCL3L correlates with diminished overall survival, although only hPCL3S levels are augmented in clinical NSCLC specimens. Inhibition of either isoform leads to reduced cell proliferation, invasion, and migration, with hPCL3S knockdown displaying superior effectiveness. Moreover, the findings reveal that TRIM21 interacts with both isoforms and mediates hPCL3S degradation through K48-linked ubiquitination in NSCLC cells. Conversely, TRIM21 does not facilitate hPCL3L degradation, despite forming K63-linked polyubiquitin chains. These observations highlight the divergent roles of hPCL3 isoforms in NSCLC and underscore the potential therapeutic value of targeting hPCL3S.