Merck
  • Plasma 11beta-hydroxy-4-androstene-3,17-dione: comparison of a time-resolved fluoroimmunoassay using a biotinylated tracer with a radioimmunoassay using a tritiated tracer.
Plasma 11beta-hydroxy-4-androstene-3,17-dione: comparison of a time-resolved fluoroimmunoassay using a biotinylated tracer with a radioimmunoassay using a tritiated tracer.
The Journal of steroid biochemistry and molecular biology (2003-05-28)
Fidaa Ibrahim, Frank Giton, Philippe Boudou, Jean Marie Villette, René Julien, Hervé Galons, Jean Fiet
ABSTRACT
The plasma concentration of 11beta-hydroxy-4-androstene-3,17-dione (11beta) is very high in 21-hydroxylase deficiency, Cushing's syndrome, and hyperandrogenism of adrenal origin, and very low in congenital 11-hydroxylase deficiency and adrenal insufficiency. Thus, when plasma 4-androstenedione is elevated, it is useful to measure the plasma 11beta level in order to determine the adrenal or ovarian origin of the hyperandrogenism. To eliminate disadvantages related to the 11beta radioimmunoassay (RIA), which uses a tritiated tracer, as well as the high cost associated with scintillation proximity assay (SPA), we developed a non-isotopic 11beta assay that utilizes an 11beta-biotin conjugate synthesized in our laboratory to measure time-resolved fluorescence after addition of streptavidin-europium to microtitration wells. The analytical qualities of this assay are very similar to those of the radioimmunoassay using a tritiated tracer, and an extraction step followed by celite chromatography (which separates 11beta from interfering plasma steroids) prior to a final radioimmuno-competition step. The correlation coefficient between 11beta levels measured by time-resolved plasma 11beta fluoroimmunoassay (TR-FIA) and RIA was 0.965.Finally, the TR-FIA technique was more sensitive and of greater precision than the RIA method.